ENA053247041 Enamine

4-(2,5-diethoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02663892
  • Cas Number : 112434-78-3
  • Molecular Weight : 264.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12961
Availability details for
Quantity
  • Availability
  • Add to Cart