ENA053247051 Enamine

3-chloro-4-methyl-5-nitrobenzenesulfonyl chloride

  • MDL number MFCD06655594
  • Cas Number : 78726-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12971

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12971
Availability details for
Quantity
  • Availability
  • Add to Cart