ENA053247058 Enamine

6-hydrazinyl-8-oxa-3,5-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),3,5,10,12-hexaene

  • MDL number MFCD00452697
  • Cas Number : 65024-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12978
Availability details for
Quantity
  • Availability
  • Add to Cart