ENA053247061 Enamine

2-[(2,4-dichlorobenzyl)amino]butan-1-ol

  • MDL number MFCD04478126
  • Cas Number : 852840-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12981
Availability details for
Quantity
  • Availability
  • Add to Cart