ENA053247062 Enamine

3-methyl-5-(propan-2-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD06655598
  • Cas Number : 90087-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-152
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12982
Availability details for
Quantity
  • Availability
  • Add to Cart