ENA053247070 Enamine

3-[(4-fluorophenyl)methoxy]benzoic acid

  • MDL number MFCD04536523
  • Cas Number : 457-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-149
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12990
Availability details for
Quantity
  • Availability
  • Add to Cart