ENA053247090 Enamine

3-nitro-4-(phenylamino)benzoic acid

  • MDL number MFCD00437310
  • Cas Number : 16927-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13010
Availability details for
Quantity
  • Availability
  • Add to Cart