ENA053247091 Enamine

2-[({4-oxo-5-phenyl-3H,4H-thieno[2,3-d]pyrimidin-2-yl}methyl)sulfanyl]propanoic acid

  • MDL number MFCD06655608
  • Cas Number : 852933-47-2
  • Molecular Weight : 346.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13011
Availability details for
Quantity
  • Availability
  • Add to Cart