ENA053247096 Enamine

1-(4-methylphenyl)imidazolidine-2,4,5-trione

  • MDL number MFCD06655609
  • Cas Number : 93284-13-0
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13016
Availability details for
Quantity
  • Availability
  • Add to Cart