ENA053247100 Enamine

4-chloro-3-(cyclopropylsulfamoyl)benzoic acid

  • MDL number MFCD06655611
  • Cas Number : 852933-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13020
Availability details for
Quantity
  • Availability
  • Add to Cart