ENA053247101 Enamine

5-methyl-6-phenyl-3-(prop-2-en-1-yl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06655612
  • Cas Number : 852933-46-1
  • Molecular Weight : 314.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13021
Availability details for
Quantity
  • Availability
  • Add to Cart