ENA053247109 Enamine

N-benzyl-N-(2,2,2-trifluoroethyl)amine

  • MDL number MFCD02689998
  • Cas Number : 85963-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13029
Availability details for
Quantity
  • Availability
  • Add to Cart