ENA053247115 Enamine

2-hydrazino-3-(3-methoxypropyl)quinazolin-4(3H)-one

  • MDL number MFCD06655616
  • Cas Number : 852933-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13035

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13035
Availability details for
Quantity
  • Availability
  • Add to Cart