ENA053247123 Enamine

5-{[(thiophen-2-yl)methyl]sulfanyl}-1,3,4-thiadiazol-2-amine

  • MDL number MFCD06655622
  • Cas Number : 852933-95-0
  • Molecular Weight : 229.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13043
Availability details for
Quantity
  • Availability
  • Add to Cart