ENA053247126 Enamine

1-(4-methoxyphenyl)-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD03848152
  • Cas Number : 3807-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13046
Availability details for
Quantity
  • Availability
  • Add to Cart