ENA053247133 Enamine

2-chloro-N-(cyclohex-1-en-1-yl)-N-[(furan-2-yl)methyl]acetamide

  • MDL number MFCD06655628
  • Cas Number : 69763-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13053
Availability details for
Quantity
  • Availability
  • Add to Cart