ENA053247147 Enamine

1,2-bis(4-methoxyphenyl)ethan-1-amine

  • MDL number MFCD06655633
  • Cas Number : 36265-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13067
Availability details for
Quantity
  • Availability
  • Add to Cart