ENA053247151 Enamine

N-{3-[4-(2-chloroacetyl)phenyl]propyl}acetamide

  • MDL number MFCD06655636
  • Cas Number : 852851-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13071

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13071
Availability details for
Quantity
  • Availability
  • Add to Cart