ENA053247154 Enamine

N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]-3-methylbutanamide

  • MDL number MFCD05511014
  • Cas Number : 852851-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13074

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13074
Availability details for
Quantity
  • Availability
  • Add to Cart