ENA053247155 Enamine

2-{[(4-methylphenyl)methyl]sulfanyl}ethan-1-amine

  • MDL number MFCD06655639
  • Cas Number : 106670-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13075
Availability details for
Quantity
  • Availability
  • Add to Cart