ENA053247159 Enamine

2-{[(2-fluorophenyl)methyl]sulfanyl}ethan-1-amine

  • MDL number MFCD06655642
  • Cas Number : 753394-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13079
Availability details for
Quantity
  • Availability
  • Add to Cart