ENA053247165 Enamine

[1-(4-fluorophenyl)-5-oxo-2,5-dihydro-1H-pyrazol-3-yl]acetic acid

  • MDL number MFCD06655644
  • Cas Number : 852851-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13085

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13085
Availability details for
Quantity
  • Availability
  • Add to Cart