ENA053247167 Enamine

2-[1-(2-methylpropyl)-5-oxo-2,5-dihydro-1H-pyrazol-3-yl]acetohydrazide

  • MDL number MFCD06655646
  • Cas Number : 852851-72-0
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13087
Availability details for
Quantity
  • Availability
  • Add to Cart