ENA053247170 Enamine

methyl 2-(5-oxo-1-phenyl-2,5-dihydro-1H-pyrazol-3-yl)acetate

  • MDL number MFCD06655649
  • Cas Number : 105041-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13090
Availability details for
Quantity
  • Availability
  • Add to Cart