ENA053247171 Enamine

(5-oxo-1-phenyl-2,5-dihydro-1H-pyrazol-3-yl)acetic acid

  • MDL number MFCD00453404
  • Cas Number : 37959-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13091

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13091
Availability details for
Quantity
  • Availability
  • Add to Cart