ENA053247173 Enamine

methyl 2-(1-tert-butyl-5-oxo-2,5-dihydro-1H-pyrazol-3-yl)acetate

  • MDL number MFCD06655652
  • Cas Number : 852851-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13093
Availability details for
Quantity
  • Availability
  • Add to Cart