ENA053247184 Enamine

5-[(diethylamino)sulfonyl]-2-[(2-hydroxyethyl)amino]benzoic acid

  • MDL number MFCD06655662
  • Cas Number : 852851-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13104

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13104
Availability details for
Quantity
  • Availability
  • Add to Cart