ENA053247190 Enamine

5-(pyrrolidine-1-sulfonyl)-1,3-benzoxazol-2-amine

  • MDL number MFCD06655667
  • Cas Number : 852851-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13110
Availability details for
Quantity
  • Availability
  • Add to Cart