ENA053247191 Enamine

5-(ethanesulfonyl)-1,3-benzoxazol-2-amine

  • MDL number MFCD06655668
  • Cas Number : 852851-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-144
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13111
Availability details for
Quantity
  • Availability
  • Add to Cart