ENA053247202 Enamine

9-chloro-3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde

  • MDL number MFCD06655675
  • Cas Number : 852851-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13122
Availability details for
Quantity
  • Availability
  • Add to Cart