ENA053247206 Enamine

2-chloro-1-(5-methylthiophen-2-yl)ethan-1-one

  • MDL number MFCD00464874
  • Cas Number : 31772-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13126
Availability details for
Quantity
  • Availability
  • Add to Cart