ENA053247210 Enamine

4-chloro-2-(piperidine-1-carbonyl)pyridine

  • MDL number MFCD06655679
  • Cas Number : 852851-85-5
  • Molecular Weight : 224.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13130
Availability details for
Quantity
  • Availability
  • Add to Cart