ENA053247212 Enamine

1-{[4-(dimethylamino)phenyl]methyl}-1H-pyrazol-5-amine

  • MDL number MFCD03596778
  • Cas Number : 3524-27-4
  • Molecular Weight : 216.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-61
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13132
Availability details for
Quantity
  • Availability
  • Add to Cart