ENA053247214 Enamine

1-(4-methoxyphenyl)imidazolidine-2,4,5-trione

  • MDL number MFCD06655682
  • Cas Number : 852956-32-2
  • Molecular Weight : 220.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13134
Availability details for
Quantity
  • Availability
  • Add to Cart