ENA053247219 Enamine

6-methyl-1-phenyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidine-4-thione

  • MDL number MFCD01847773
  • Cas Number : 5399-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13148
Availability details for
Quantity
  • Availability
  • Add to Cart