ENA053247222 Enamine

2-(tert-butylsulfanyl)acetic acid

  • MDL number MFCD06655686
  • Cas Number : 24310-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13151
Availability details for
Quantity
  • Availability
  • Add to Cart