ENA053247229 Enamine

N-cyclopropylurea

  • MDL number MFCD01569844
  • Cas Number : 19757-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13158

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13158
Availability details for
Quantity
  • Availability
  • Add to Cart