ENA053247230 Enamine

3-(carbamoylamino)benzoic acid

  • MDL number MFCD02379821
  • Cas Number : 20632-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265-270
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13159
Availability details for
Quantity
  • Availability
  • Add to Cart