ENA053247239 Enamine

3-(1-isopropyl-4,6-dimethyl-1H-pyrazolo[3,4-b]pyridin-5-yl)propanoic acid

  • MDL number MFCD06655695
  • Cas Number : 853574-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13168
Availability details for
Quantity
  • Availability
  • Add to Cart