ENA053247244 Enamine

methyl({1-[4-(1H-1,2,4-triazol-1-yl)phenyl]ethyl})amine

  • MDL number MFCD06655700
  • Cas Number : 853574-50-2
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13173
Availability details for
Quantity
  • Availability
  • Add to Cart