ENA053247245 Enamine

4-(5-ethylthiophen-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD02090887
  • Cas Number : 524932-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13174
Availability details for
Quantity
  • Availability
  • Add to Cart