ENA053247247 Enamine

1-(5-ethylthiophen-2-yl)ethan-1-one

  • MDL number MFCD00130088
  • Cas Number : 18761-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13176
Availability details for
Quantity
  • Availability
  • Add to Cart