ENA053247257 Enamine

2-(2-tert-butylphenoxy)acetic acid

  • MDL number MFCD06655706
  • Cas Number : 19271-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13186
Availability details for
Quantity
  • Availability
  • Add to Cart