ENA053247259 Enamine

1-bromo-2-(chloromethyl)-4,5-dimethoxybenzene

  • MDL number MFCD06655708
  • Cas Number : 54370-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13188

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13188
Availability details for
Quantity
  • Availability
  • Add to Cart