ENA053247265 Enamine

2-(allylthio)aniline

  • MDL number MFCD06655714
  • Cas Number : 77053-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13194

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13194
Availability details for
Quantity
  • Availability
  • Add to Cart