ENA053247266 Enamine

2-chloro-1-(5-ethylthiophen-2-yl)ethan-1-one

  • MDL number MFCD06655715
  • Cas Number : 72612-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13195
Availability details for
Quantity
  • Availability
  • Add to Cart