ENA053247268 Enamine

2-chloro-N-(2-{[(4-chlorophenyl)methyl]sulfanyl}ethyl)acetamide

  • MDL number MFCD06655717
  • Cas Number : 853574-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13197
Availability details for
Quantity
  • Availability
  • Add to Cart