ENA053247269 Enamine

2-{[2-(4-chlorophenyl)quinolin-4-yl]formamido}acetic acid

  • MDL number MFCD02711461
  • Cas Number : 325720-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13198
Availability details for
Quantity
  • Availability
  • Add to Cart