ENA053247272 Enamine

5-[(4-methylpiperazin-1-yl)sulfonyl]pyridine-2-thiol

  • MDL number MFCD06655719
  • Cas Number : 853574-40-0
  • Molecular Weight : 273.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13201
Availability details for
Quantity
  • Availability
  • Add to Cart