ENA053247280 Enamine

2-[(2,4-dichlorobenzoyl)amino]-3-phenylpropanoic acid

  • MDL number MFCD06655722
  • Cas Number : 1396980-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13209
Availability details for
Quantity
  • Availability
  • Add to Cart